Product Details

SNP ID
rs17641143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:45510227 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCCCGGATTTTGGAAGGACGCGG[C/T]CTCACAATAAAAGACAGTATTGTAG
Phenotype
MIM: 135820
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBLN1 PubMed Links

Gene Details

Gene
FBLN1
Gene Name
fibulin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001996.3 Intron NP_001987.2
NM_006485.3 Intron NP_006476.2
NM_006486.2 Intron NP_006477.2
NM_006487.2 Intron NP_006478.2

View Full Product Details