Product Details
- SNP ID
-
rs17092915
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:35651548 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTTAAAAACCTAGATATAACACT[C/T]TCTTATGAATGGGCCCCTCTTTGTT
- Phenotype
-
MIM: 604205
MIM: 607179
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CPNE1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76401607] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPNE1
- Gene Name
- copine 1
- Gene
- RBM12
- Gene Name
- RNA binding motif protein 12
View Full Product Details