Product Details

SNP ID
rs17524694
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:51450989 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTACTTCATTTAACCAAACTTA[C/T]TTGAGCATGGAGCACTTTTTGGGTG
Phenotype
MIM: 612186
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DMXL2 PubMed Links

Gene Details

Gene
DMXL2
Gene Name
Dmx like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174116.1 Intron NP_001167587.1
NM_001174117.1 Intron NP_001167588.1
NM_015263.3 Intron NP_056078.2
XM_005254255.1 Intron XP_005254312.1
XM_005254256.1 Intron XP_005254313.1
XM_011521402.2 Intron XP_011519704.1
XM_017022034.1 Intron XP_016877523.1

View Full Product Details