Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001144955.1 | 430 | Intron | NP_001138427.1 | ||
NM_020234.5 | 430 | Intron | NP_064619.2 | ||
XM_011521815.1 | 430 | Intron | XP_011520117.1 | ||
XM_017022419.1 | 430 | Intron | XP_016877908.1 | ||
XM_017022420.1 | 430 | UTR 5 | XP_016877909.1 | ||
XM_017022421.1 | 430 | Intron | XP_016877910.1 | ||
XM_017022422.1 | 430 | UTR 5 | XP_016877911.1 | ||
XM_017022423.1 | 430 | Intron | XP_016877912.1 | ||
XM_017022424.1 | 430 | Intron | XP_016877913.1 | ||
XM_017022425.1 | 430 | Intron | XP_016877914.1 | ||
XM_017022426.1 | 430 | UTR 5 | XP_016877915.1 | ||
XM_017022427.1 | 430 | Intron | XP_016877916.1 | ||
XM_017022428.1 | 430 | Intron | XP_016877917.1 | ||
XM_017022429.1 | 430 | UTR 5 | XP_016877918.1 | ||
XM_017022430.1 | 430 | Intron | XP_016877919.1 | ||
XM_017022431.1 | 430 | Intron | XP_016877920.1 |