Product Details

SNP ID
rs16976367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55540264 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCAGGTAAATTAGAGGAGTCTTA[C/T]GGTATCTTAATCATGTCTAAATAAA
Phenotype
MIM: 606902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107984745 PubMed Links

Gene Details

Gene
LOC107984745
Gene Name
uncharacterized mitochondrial protein AtMg00860-like
There are no transcripts associated with this gene.

Gene
PYGO1
Gene Name
pygopus family PHD finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015617.2 7414 UTR 3 NP_056432.1
XM_005254300.4 7414 UTR 3 XP_005254357.1
XM_011521446.2 7414 UTR 3 XP_011519748.1

View Full Product Details