Product Details

SNP ID
rs17799519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90873116 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAACTCATCTTATACTATACTTCA[A/C]ATGAATTATCTTAATATATTTGCAT
Phenotype
MIM: 603607
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPS6KA5 PubMed Links

Gene Details

Gene
RPS6KA5
Gene Name
ribosomal protein S6 kinase A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322227.1 Intron NP_001309156.1
NM_001322228.1 Intron NP_001309157.1
NM_001322229.1 Intron NP_001309158.1
NM_001322230.1 Intron NP_001309159.1
NM_001322231.1 Intron NP_001309160.1
NM_001322232.1 Intron NP_001309161.1
NM_001322233.1 Intron NP_001309162.1
NM_001322234.1 Intron NP_001309163.1
NM_001322235.1 Intron NP_001309164.1
NM_001322236.1 Intron NP_001309165.1
NM_001322237.1 Intron NP_001309166.1
NM_001322238.1 Intron NP_001309167.1
NM_004755.3 Intron NP_004746.2
NM_182398.2 Intron NP_872198.1
XM_017021786.1 Intron XP_016877275.1

View Full Product Details