Product Details

SNP ID
rs17154400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107748914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAAGGCACTAATAGTTCTGGATC[C/T]TTGGGTGGTCTTGATGTTCGCAGAC
Phenotype
MIM: 606872
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBLL1 PubMed Links

Gene Details

Gene
CBLL1
Gene Name
Cbl proto-oncogene like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284291.1 316 Silent Mutation TCC,TCT S16S NP_001271220.1
NM_024814.3 316 Silent Mutation TCC,TCT S16S NP_079090.2
XM_011516580.2 316 UTR 5 XP_011514882.1
XM_017012643.1 316 Silent Mutation TCC,TCT S56S XP_016868132.1
XM_017012644.1 316 UTR 5 XP_016868133.1
XM_017012645.1 316 UTR 5 XP_016868134.1

View Full Product Details