Product Details

SNP ID
rs16997503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21187827 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGTATGCATCACAGCTATTCTTT[C/T]TCTGAGTTATAGCTACAGTTTTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF431 PubMed Links

Gene Details

Gene
ZNF431
Gene Name
zinc finger protein 431
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319124.1 Intron NP_001306053.1
NM_001319126.1 Intron NP_001306055.1
NM_001319127.1 Intron NP_001306056.1
XM_011527750.2 Intron XP_011526052.1

View Full Product Details