Product Details

SNP ID
rs118203933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:85473779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACTTGATGGACAAGGTTCAGAG[C/T]ATACTGTGGATAAAAAGAAATATGC
Phenotype
MIM: 611492
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CA2 PubMed Links

Gene Details

Gene
CA2
Gene Name
carbonic anhydrase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000067.2 499 Missense Mutation CAT,TAT H107Y NP_000058.1
NM_001293675.1 499 Intron NP_001280604.1
Gene
CA3-AS1
Gene Name
CA3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details