Product Details

SNP ID
rs17227152
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:964950 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACATGACAGATGATGAACTACCC[A/G]AAATATCTCGTGAGAAGCTCTGGCA
Phenotype
MIM: 608114 MIM: 610429
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ADAP1 PubMed Links

Gene Details

Gene
ADAP1
Gene Name
ArfGAP with dual PH domains 1
There are no transcripts associated with this gene.

Gene
COX19
Gene Name
COX19, cytochrome c oxidase assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031617.2 4791 UTR 3 NP_001026788.1

View Full Product Details