Product Details

SNP ID
rs17661499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:13494573 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATATTATTTTAGAGTGTCTGGTC[A/G]TTTAAATTTTGGCAGATAAAGAAAA
Phenotype
MIM: 604057
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HS3ST3A1 PubMed Links

Gene Details

Gene
HS3ST3A1
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 3A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006042.2 3658 UTR 3 NP_006033.1
XM_011524114.2 3658 UTR 3 XP_011522416.1
XM_017025480.1 3658 Intron XP_016880969.1

View Full Product Details