Product Details

SNP ID
rs17345517
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:169367882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCTGGGAGTAATCTCAGCTGC[C/G]TTCATCCAATCGACAGACCCGCCCC
Phenotype
MIM: 608692 MIM: 613465
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BLZF1 PubMed Links

Gene Details

Gene
BLZF1
Gene Name
basic leucine zipper nuclear factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320972.1 68 UTR 5 NP_001307901.1
NM_001320973.1 68 UTR 5 NP_001307902.1
NM_003666.3 68 UTR 5 NP_003657.1
XM_005245561.4 68 Intron XP_005245618.1
XM_011510092.2 68 Intron XP_011508394.1
XM_017002645.1 68 Intron XP_016858134.1
Gene
NME7
Gene Name
NME/NM23 family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013330.4 68 UTR 5 NP_037462.1
NM_197972.2 68 UTR 5 NP_932076.1
XM_005245106.4 68 Intron XP_005245163.1
XM_017001099.1 68 UTR 5 XP_016856588.1
XM_017001100.1 68 Intron XP_016856589.1

View Full Product Details