Product Details

SNP ID
rs16872639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75623031 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAATTACCTTTTAGGGAAAAATA[G/T]ATCTCCTATCAAAGAGTGATTTGTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKDD1B PubMed Links

Gene Details

Gene
ANKDD1B
Gene Name
ankyrin repeat and death domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276713.1 Intron NP_001263642.1
XM_011543618.2 Intron XP_011541920.1
XM_011543623.2 Intron XP_011541925.1
XM_017009814.1 Intron XP_016865303.1
XM_017009815.1 Intron XP_016865304.1
XM_017009816.1 Intron XP_016865305.1
XM_017009817.1 Intron XP_016865306.1

View Full Product Details