Product Details

SNP ID
rs17432206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55242794 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGCACATCCTATTAAGTTTCAAT[C/T]GCTAGCTGGGACTGGACTATAGCAG
Phenotype
MIM: 603766 MIM: 191343
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTIF2 PubMed Links

Gene Details

Gene
MTIF2
Gene Name
mitochondrial translational initiation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005369.1 Intron NP_001005369.1
NM_001321001.1 Intron NP_001307930.1
NM_001321002.1 Intron NP_001307931.1
NM_001321003.1 Intron NP_001307932.1
NM_001321004.1 Intron NP_001307933.1
NM_001321005.1 Intron NP_001307934.1
NM_002453.2 Intron NP_002444.2
XM_005264335.3 Intron XP_005264392.1
XM_011532871.2 Intron XP_011531173.1
XM_017004162.1 Intron XP_016859651.1
XM_017004163.1 Intron XP_016859652.1
XM_017004164.1 Intron XP_016859653.1
XM_017004165.1 Intron XP_016859654.1
XM_017004166.1 Intron XP_016859655.1
XM_017004167.1 Intron XP_016859656.1
Gene
RPS27A
Gene Name
ribosomal protein S27a
There are no transcripts associated with this gene.

View Full Product Details