Product Details

SNP ID
rs17705020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35841999 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACTGGCAGCCTAGGCTTTTCCCA[C/T]TGGCTACCACAACATGTAAATCCAT
Phenotype
MIM: 601939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRPK1 PubMed Links

Gene Details

Gene
SRPK1
Gene Name
SRSF protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003137.4 Intron NP_003128.3

View Full Product Details