Product Details

SNP ID
rs7663617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:75725588 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACGTCCACCGCTTTCTGCCTGAA[C/T]TCCTAATGACATGATTTGCGGCGTT
Phenotype
MIM: 603344
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
G3BP2 PubMed Links

Gene Details

Gene
G3BP2
Gene Name
G3BP stress granule assembly factor 2
There are no transcripts associated with this gene.

Gene
USO1
Gene Name
USO1 vesicle transport factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290049.1 Intron NP_001276978.1
NM_003715.3 Intron NP_003706.2
XM_006714396.3 Intron XP_006714459.1
XM_006714397.3 Intron XP_006714460.1

View Full Product Details