Product Details

SNP ID
rs17585970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109563187 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAAATGTGCCTTTCTTAATTTGGA[C/T]TTTGTGCTACTTAGACTGAGAAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCUB PubMed Links

Gene Details

Gene
MCUB
Gene Name
mitochondrial calcium uniporter dominant negative beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017918.4 Intron NP_060388.2
XM_006714246.3 Intron XP_006714309.1

View Full Product Details