Product Details

SNP ID
rs17063109
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:133891432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGCTTGCCCGGGGCTTCCTGCA[C/T]TGAGTCTCACAAGGAACCCAGTATA
Phenotype
MIM: 616058 MIM: 603306
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TARID PubMed Links
Additional Information
For this assay, SNP(s) [rs4896010] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
TARID
Gene Name
TCF21 antisense RNA inducing promoter demethylation
There are no transcripts associated with this gene.

Gene
TCF21
Gene Name
transcription factor 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003206.3 Intron NP_003197.2
NM_198392.2 Intron NP_938206.1

View Full Product Details