Product Details
- SNP ID
-
rs17459137
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:115267758 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTTAATACCTCTCAATTCACAATA[A/T]AGCACAATAATCAATCTGACTAAGC
- Phenotype
-
MIM: 602031
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC112
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75094465] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC112
- Gene Name
- coiled-coil domain containing 112
- Gene
- PGGT1B
- Gene Name
- protein geranylgeranyltransferase type I subunit beta
There are no transcripts associated with this gene.
View Full Product Details