Product Details
- SNP ID
-
rs17220099
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:130206335 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATATTTCACCTCTATAGGAAACCA[C/G]TGCATACCTTGGATTGACTTCTTTA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF280C
PubMed Links
Gene Details
- Gene
- ZNF280C
- Gene Name
- zinc finger protein 280C
View Full Product Details