Product Details

SNP ID
rs59715127
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.7:99757867 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
AGGAGGAGTTAATGGTGCTAACTGG[G/T]GGTGGTGGAAATAGTCCCGTGAGAA
Phenotype
MIM: 124010
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
CYP3A4*1H,g.26206C>A
Literature Links
CYP3A4 PubMed Links

Gene Details

Gene
CYP3A4
Gene Name
cytochrome P450 family 3 subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202855.2 1882 UTR 3 NP_001189784.1
NM_017460.5 1882 UTR 3 NP_059488.2
XM_017011780.1 1882 Intron XP_016867269.1

View Full Product Details