Product Details
- SNP ID
-
rs1042881
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:161526740 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATAACTGAAGTCTTTTGACTTTTTG[C/G]GGGGAGGGCGGTTCATCCTCTTCTG
- Phenotype
-
MIM: 146790
MIM: 140555
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FCGR2A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41299264] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FCGR2A
- Gene Name
- Fc fragment of IgG receptor IIa
There are no transcripts associated with this gene.
- Gene
- HSPA6
- Gene Name
- heat shock protein family A (Hsp70) member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002155.4 |
2201 |
UTR 3 |
|
|
NP_002146.2 |
View Full Product Details