Product Details

SNP ID
rs6724844
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:131484879 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCTGAACACCAGAGGCTCTCTC[A/G]GATGAAGACCACAGGTTCAGAGTTT
Phenotype
MIM: 613449
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC150776 PubMed Links
Additional Information
For this assay, SNP(s) [rs200606526] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC150776
Gene Name
sphingomyelin phosphodiesterase 4, neutral membrane (neutral sphingomyelinase-3) pseudogene
There are no transcripts associated with this gene.

Gene
MIR4784
Gene Name
microRNA 4784
There are no transcripts associated with this gene.

Gene
MZT2A
Gene Name
mitotic spindle organizing protein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085365.1 Intron NP_001078834.1
XM_005263741.4 Intron XP_005263798.1
XM_005263742.2 Intron XP_005263799.1
XM_017004744.1 Intron XP_016860233.1
Gene
POTEI
Gene Name
POTE ankyrin domain family member I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277406.1 Intron NP_001264335.1
XM_011511670.2 Intron XP_011509972.1
XM_017004732.1 Intron XP_016860221.1
XM_017004733.1 Intron XP_016860222.1
XM_017004734.1 Intron XP_016860223.1
XM_017004735.1 Intron XP_016860224.1
Gene
TUBA3D
Gene Name
tubulin alpha 3d
There are no transcripts associated with this gene.

View Full Product Details