Product Details

SNP ID
rs11556469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240587010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTCTTCTGCGACTTGTCTACGCC[G/T]CTGGCCCAGTTCCGCGAGGACATCA
Phenotype
MIM: 605286 MIM: 605287
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CAPN10 PubMed Links

Gene Details

Gene
CAPN10
Gene Name
calpain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023083.3 295 Silent Mutation CCG,CCT P33P NP_075571.1
NM_023085.3 295 Silent Mutation CCG,CCT P33P NP_075573.2
Gene
CAPN10-AS1
Gene Name
CAPN10 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
RNPEPL1
Gene Name
arginyl aminopeptidase like 1
There are no transcripts associated with this gene.

View Full Product Details