Product Details

SNP ID
rs13419754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:239894893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGCTACAGGCACGATCCTGAGG[C/T]ACCTCAAATTTAGCAGGTCCCAAGT
Phenotype
MIM: 603835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFA10 PubMed Links

Gene Details

Gene
NDUFA10
Gene Name
NADH:ubiquinone oxidoreductase subunit A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322019.1 Intron NP_001308948.1
NM_001322020.1 Intron NP_001308949.1
NM_004544.3 Intron NP_004535.1
XM_011511228.2 Intron XP_011509530.1

View Full Product Details