Product Details

SNP ID
rs2972191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73447055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCAAGGCGGCGGACGAGGAGAC[C/T]GAGGAGGCGGCGGCGGCCGCGGCGG
Phenotype
MIM: 601091
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXD1 PubMed Links

Gene Details

Gene
FOXD1
Gene Name
forkhead box D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004472.2 1473 Silent Mutation TCA,TCG S436S NP_004463.1
Gene
LINC01386
Gene Name
long intergenic non-protein coding RNA 1386
There are no transcripts associated with this gene.

View Full Product Details