Product Details

SNP ID
rs7349803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80417274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCCAAAGGTCAGCACCTTATTC[C/T]TACGTCCCCTGTGATGAGGTTGTTT
Phenotype
MIM: 608880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFYVE16 PubMed Links

Gene Details

Gene
ZFYVE16
Gene Name
zinc finger FYVE-type containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105251.2 Intron NP_001098721.1
NM_001284236.1 Intron NP_001271165.1
NM_001284237.1 Intron NP_001271166.1
NM_014733.4 Intron NP_055548.3
XM_005248632.4 Intron XP_005248689.1
XM_011543753.2 Intron XP_011542055.1
XM_017010091.1 Intron XP_016865580.1
XM_017010092.1 Intron XP_016865581.1
XM_017010093.1 Intron XP_016865582.1
XM_017010094.1 Intron XP_016865583.1
XM_017010095.1 Intron XP_016865584.1
XM_017010096.1 Intron XP_016865585.1

View Full Product Details