Product Details

SNP ID
rs11551763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89105197 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCGAGTTCTTCTTTGGCTGGGGCTT[A/C]TTGATCTCCTGTGATCACTTGGAGA
Phenotype
MIM: 615913
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PM20D2 PubMed Links

Gene Details

Gene
PM20D2
Gene Name
peptidase M20 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010853.2 788 Intron NP_001010853.1
XM_005248661.2 788 Intron XP_005248718.1
XM_005248662.4 788 Intron XP_005248719.1
XM_011535481.2 788 Intron XP_011533783.1
XM_017010289.1 788 Intron XP_016865778.1
XM_017010290.1 788 Intron XP_016865779.1
XM_017010291.1 788 Intron XP_016865780.1
Gene
SRSF12
Gene Name
serine and arginine rich splicing factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080743.4 788 Missense Mutation AGA,ATA R113I NP_542781.3
XM_006715348.2 788 Missense Mutation AGA,ATA R18I XP_006715411.1
XM_011535483.2 788 Missense Mutation AGA,ATA R98I XP_011533785.1
XM_011535484.1 788 Missense Mutation AGA,ATA R18I XP_011533786.1
XM_017010292.1 788 Missense Mutation AGA,ATA R113I XP_016865781.1
XM_017010293.1 788 Missense Mutation AGA,ATA R18I XP_016865782.1

View Full Product Details