Product Details

SNP ID
rs7769545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28271797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTATAAACCCAAGAAAATGTTA[C/T]TGTTCTTGTTAGCAGTACTATTACT
Phenotype
MIM: 616474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGBD1 PubMed Links

Gene Details

Gene
PGBD1
Gene Name
piggyBac transposable element derived 1
There are no transcripts associated with this gene.

Gene
ZSCAN26
Gene Name
zinc finger and SCAN domain containing 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023560.3 Intron NP_001018854.2
NM_001111039.2 Intron NP_001104509.1
NM_001287421.1 Intron NP_001274350.1
NM_001287422.1 Intron NP_001274351.1
NM_152736.5 Intron NP_689949.3
XM_011514862.2 Intron XP_011513164.1
XM_011514864.1 Intron XP_011513166.1
XM_011514866.1 Intron XP_011513168.1
XM_011514867.1 Intron XP_011513169.1
XM_011514869.1 Intron XP_011513171.1
XM_017011264.1 Intron XP_016866753.1
XM_017011265.1 Intron XP_016866754.1

View Full Product Details