Product Details

SNP ID
rs6963446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150719418 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGTAGAATGTTTAGAGAACAAGG[A/G]TAGAGGGTTAAAACATATATTCTTT
Phenotype
MIM: 608084
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIMAP1 PubMed Links

Gene Details

Gene
GIMAP1
Gene Name
GTPase, IMAP family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130759.3 Intron NP_570115.1
Gene
GIMAP1-GIMAP5
Gene Name
GIMAP1-GIMAP5 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199577.1 Intron NP_001186506.1
NM_001303630.1 Intron NP_001290559.1

View Full Product Details