Product Details

SNP ID
rs11551220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:40153795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGATGGCCTTGAAGAAGAGGA[A/C]AAAAGACAAGCTTTTCCAGTTTCTG
Phenotype
MIM: 607702
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C8orf4 PubMed Links

Gene Details

Gene
C8orf4
Gene Name
chromosome 8 open reading frame 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020130.4 328 Missense Mutation AAA,ACA K88T NP_064515.1

View Full Product Details