Product Details

SNP ID
rs7007344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:48728494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCTACACATATCTCTATGACTC[C/T]GGCACCAGGGTACCCAGGGTATTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EFCAB1 PubMed Links

Gene Details

Gene
EFCAB1
Gene Name
EF-hand calcium binding domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142857.1 Intron NP_001136329.1
NM_024593.3 Intron NP_078869.1
XM_005251303.1 Intron XP_005251360.1
XM_011517589.1 Intron XP_011515891.1
XM_011517590.2 Intron XP_011515892.1
XM_011517591.2 Intron XP_011515893.1
XM_017013826.1 Intron XP_016869315.1
XM_017013827.1 Intron XP_016869316.1
XM_017013828.1 Intron XP_016869317.1

View Full Product Details