Product Details

SNP ID
rs7813634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81705614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTCATCCACAATACCGCATAAA[A/G]AGACATAAGATAGAAACAATTATAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFAND1 PubMed Links

Gene Details

Gene
ZFAND1
Gene Name
zinc finger AN1-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170796.1 Intron NP_001164267.1
NM_001170797.1 Intron NP_001164268.1
NM_024699.2 Intron NP_078975.2

View Full Product Details