Product Details

SNP ID
rs7875340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37610105 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTGAACATACTACATAGTTTTCT[A/T]AATTTGTTGTCTCTTTCCCCACAAT
Phenotype
MIM: 616919
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FRMPD1 PubMed Links

Gene Details

Gene
FRMPD1
Gene Name
FERM and PDZ domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014907.2 Intron NP_055722.2
XM_011517803.2 Intron XP_011516105.1
XM_011517804.2 Intron XP_011516106.1
XM_011517805.2 Intron XP_011516107.1
XM_011517806.2 Intron XP_011516108.1
XM_017014480.1 Intron XP_016869969.1
XM_017014481.1 Intron XP_016869970.1
XM_017014482.1 Intron XP_016869971.1

View Full Product Details