Product Details

SNP ID
rs7860058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122546032 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTATGTTAGGCACGTGTGTGTT[A/G]GGGGGTGGATAAACTTGTCGCTTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1L8 PubMed Links

Gene Details

Gene
OR1L8
Gene Name
olfactory receptor family 1 subfamily L member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004454.1 1986 Intron NP_001004454.1
XM_017014284.1 1986 Intron XP_016869773.1
XM_017014285.1 1986 UTR 3 XP_016869774.1
XM_017014286.1 1986 Intron XP_016869775.1
Gene
OR1N2
Gene Name
olfactory receptor family 1 subfamily N member 2
There are no transcripts associated with this gene.

View Full Product Details