Product Details

SNP ID
rs7854499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128085662 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTTCTGATCTCTTAGGGTGGAA[A/G]CTGAAGTCACTGATTTTGAGACCTT
Phenotype
MIM: 608745
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A25 PubMed Links

Gene Details

Gene
SLC25A25
Gene Name
solute carrier family 25 member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006641.3 Intron NP_001006642.1
NM_001006642.3 Intron NP_001006643.1
NM_001265614.2 Intron NP_001252543.1
NM_052901.4 Intron NP_443133.2
XM_005251688.1 Intron XP_005251745.1
XM_005251689.4 Intron XP_005251746.1
XM_006716948.2 Intron XP_006717011.1
XM_006716949.3 Intron XP_006717012.1

View Full Product Details