Product Details
- SNP ID
-
rs11549260
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128166354 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCGTTCCGGGCGTTGATTGCGCAC[C/T]GGCGGCTCACAGGTCGGGTGGTGGG
- Phenotype
-
MIM: 611420
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C9orf16
PubMed Links
Gene Details
- Gene
- C9orf16
- Gene Name
- chromosome 9 open reading frame 16
There are no transcripts associated with this gene.
- Gene
- CIZ1
- Gene Name
- CDKN1A interacting zinc finger protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001131015.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q791R |
NP_001124487.1 |
NM_001131016.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q847R |
NP_001124488.1 |
NM_001131017.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q786R |
NP_001124489.1 |
NM_001131018.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q767R |
NP_001124490.1 |
NM_001257975.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q903R |
NP_001244904.1 |
NM_001257976.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q746R |
NP_001244905.1 |
NM_012127.2 |
2743 |
Missense Mutation |
CAG,CGG |
Q847R |
NP_036259.2 |
XM_005251888.3 |
2743 |
Missense Mutation |
CAG,CGG |
Q847R |
XP_005251945.2 |
XM_005251891.3 |
2743 |
Missense Mutation |
CAG,CGG |
Q791R |
XP_005251948.2 |
XM_005251892.3 |
2743 |
Missense Mutation |
CAG,CGG |
Q786R |
XP_005251949.2 |
XM_005251893.4 |
2743 |
Missense Mutation |
CAG,CGG |
Q767R |
XP_005251950.4 |
XM_006717039.3 |
2743 |
Missense Mutation |
CAG,CGG |
Q818R |
XP_006717102.2 |
XM_011518481.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q842R |
XP_011516783.1 |
XM_011518482.2 |
2743 |
Missense Mutation |
CAG,CGG |
Q823R |
XP_011516784.2 |
XM_011518483.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q823R |
XP_011516785.1 |
XM_017014594.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q877R |
XP_016870083.1 |
XM_017014595.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q842R |
XP_016870084.1 |
XM_017014596.1 |
2743 |
Missense Mutation |
CAG,CGG |
Q791R |
XP_016870085.1 |
View Full Product Details