Product Details

SNP ID
rs7086696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70816428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGGAGGGGTGACAACGAGAGCGA[G/T]GCCGTGCTGATTCTGAAGGGAGTGC
Phenotype
MIM: 603729
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SGPL1 PubMed Links

Gene Details

Gene
SGPL1
Gene Name
sphingosine-1-phosphate lyase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003901.3 45 Intron NP_003892.2
XM_005270263.1 45 UTR 5 XP_005270320.1
XM_006718053.1 45 Intron XP_006718116.1
XM_011540316.2 45 Intron XP_011538618.1
XM_011540317.1 45 UTR 5 XP_011538619.1
XM_011540318.1 45 Intron XP_011538620.1

View Full Product Details