Product Details

SNP ID
rs7107124
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:20388053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTGTGGAGAATGAGGAGGACCT[C/G]CCAGAACTGTCGGACAGCGGGGACG
Phenotype
MIM: 605628 MIM: 603190
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
HTATIP2 PubMed Links

Gene Details

Gene
HTATIP2
Gene Name
HIV-1 Tat interactive protein 2
There are no transcripts associated with this gene.

Gene
PRMT3
Gene Name
protein arginine methyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145166.1 280 Intron NP_001138638.1
NM_001145167.1 280 Intron NP_001138639.1
NM_005788.3 280 Silent Mutation CTC,CTG L21L NP_005779.1
XM_011519836.2 280 Intron XP_011518138.1
XM_017017081.1 280 Missense Mutation TCC,TGC S5C XP_016872570.1

View Full Product Details