Product Details
- SNP ID
-
rs5030118
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:32437078 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACGCTTTGGTTTTTAGCTTGCATT[A/G]CATAAACGTTGTCGCCATTAAATTC
- Phenotype
-
MIM: 607102
MIM: 607899
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WT1
PubMed Links
Gene Details
- Gene
- WT1
- Gene Name
- Wilms tumor 1
There are no transcripts associated with this gene.
- Gene
- WT1-AS
- Gene Name
- WT1 antisense RNA
There are no transcripts associated with this gene.
View Full Product Details