Product Details

SNP ID
rs7975233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123673198 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATCAATTCTGGAGAGAAAAGGAC[A/G]GGAAAGACAAGCTCTGCTTGTACCC
Phenotype
MIM: 613846
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCTN2 PubMed Links

Gene Details

Gene
TCTN2
Gene Name
tectonic family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143850.2 Intron NP_001137322.1
NM_024809.4 Intron NP_079085.2
XM_005253623.2 Intron XP_005253680.1
XM_006719605.3 Intron XP_006719668.1
XM_017019974.1 Intron XP_016875463.1
XM_017019975.1 Intron XP_016875464.1

View Full Product Details