Product Details

SNP ID
rs7316810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111646389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATGCATCAGAACACCCAAGAG[C/G]TGAGACATGAGAAAAGAGGCACTTA
Phenotype
MIM: 604986
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BRAP PubMed Links

Gene Details

Gene
BRAP
Gene Name
BRCA1 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006768.4 Intron NP_006759.3
XM_005253944.4 Intron XP_005254001.1
XM_011538789.2 Intron XP_011537091.1
XM_017019992.1 Intron XP_016875481.1
XM_017019993.1 Intron XP_016875482.1
XM_017019994.1 Intron XP_016875483.1
XM_017019995.1 Intron XP_016875484.1

View Full Product Details