Product Details

SNP ID
rs5745796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118016864 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCACTCAAGCAGCAGGAGCAGCCC[A/G]CGGCGACCAAGATCAGGAACCTGCC
Phenotype
MIM: 600407
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RFC5 PubMed Links

Gene Details

Gene
RFC5
Gene Name
replication factor C subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130112.2 162 UTR 5 NP_001123584.1
NM_001206801.1 162 Missense Mutation ACG,GCG T13A NP_001193730.1
NM_007370.5 162 Missense Mutation ACG,GCG T13A NP_031396.1
NM_181578.3 162 UTR 5 NP_853556.2
XM_011538643.2 162 Missense Mutation ACG,GCG T13A XP_011536945.1
XM_011538645.2 162 Missense Mutation ACG,GCG T13A XP_011536947.1
XM_017019779.1 162 Missense Mutation ACG,GCG T13A XP_016875268.1

View Full Product Details