Product Details

SNP ID
rs7180081
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40036549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGTGGGAAGATGTGCAAACTGG[C/G]AGAACACCAGCCCTATCTGCATAAT
Phenotype
MIM: 609280 MIM: 600708
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
EIF2AK4 PubMed Links

Gene Details

Gene
EIF2AK4
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013703.3 Intron NP_001013725.2
XM_005254392.2 Intron XP_005254449.1
XM_011521599.2 Intron XP_011519901.1
XM_011521600.2 Intron XP_011519902.1
XM_017022219.1 Intron XP_016877708.1
Gene
SRP14
Gene Name
signal recognition particle 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309434.1 Intron NP_001296363.1
NM_003134.5 Intron NP_003125.3
Gene
SRP14-AS1
Gene Name
SRP14 antisense RNA1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details