Product Details

SNP ID
rs11075779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70689411 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAAGGAGCTCCCCCAAAACGAA[C/T]TCTTGGCCATGAGTTCAGTCCAGCA
Phenotype
MIM: 616951 MIM: 604632
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTSS1L PubMed Links

Gene Details

Gene
MTSS1L
Gene Name
metastasis suppressor 1 like
There are no transcripts associated with this gene.

Gene
VAC14
Gene Name
Vac14, PIKFYVE complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018052.3 5094 Intron NP_060522.3
XM_005256038.3 5094 UTR 3 XP_005256095.1
XM_005256041.3 5094 Intron XP_005256098.2
XM_011523223.2 5094 Intron XP_011521525.1
XM_011523224.2 5094 Intron XP_011521526.1
XM_011523225.2 5094 Intron XP_011521527.1
XM_017023441.1 5094 Intron XP_016878930.1
XM_017023442.1 5094 Intron XP_016878931.1

View Full Product Details