Product Details
- SNP ID
-
rs8062701
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89686806 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCTCCGGAACACAGGGTGCGCGG[A/G]GTGCCACCCGGGCAGCTCTGCCCGC
- Phenotype
-
MIM: 603464
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CDK10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs544949292] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDK10
- Gene Name
- cyclin dependent kinase 10
- Gene
- SPATA2L
- Gene Name
- spermatogenesis associated 2 like
There are no transcripts associated with this gene.
View Full Product Details