Product Details
- SNP ID
-
rs7225740
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:19246522 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGAGCACTCTTTGCAGCTCCCCA[A/G]CCAAGGAATGGTAAACAGCTATGTG
- Phenotype
-
MIM: 607263
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPN2
PubMed Links
Gene Details
- Gene
- EPN2
- Gene Name
- epsin 2
View Full Product Details