Product Details

SNP ID
rs8079162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43131367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAATTCGAAGTAGCGCTTAGCT[C/G]ATTTGTGTCTCAAAATAATAATAAT
Phenotype
MIM: 113705
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BRCA1 PubMed Links

Gene Details

Gene
BRCA1
Gene Name
BRCA1, DNA repair associated
There are no transcripts associated with this gene.

Gene
NBR2
Gene Name
neighbor of BRCA1 gene 2 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details