Product Details

SNP ID
rs7208227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56838968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGTTTACTTGGAAATGTACTAA[A/C]AATAAGATGAATTAATGGATAGAAA
Phenotype
MIM: 601440
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C17orf67 PubMed Links

Gene Details

Gene
C17orf67
Gene Name
chromosome 17 open reading frame 67
There are no transcripts associated with this gene.

Gene
DGKE
Gene Name
diacylglycerol kinase epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003647.2 Intron NP_003638.1
XM_011525394.2 Intron XP_011523696.1
XM_011525395.2 Intron XP_011523697.1
XM_011525396.2 Intron XP_011523698.1
XM_017025243.1 Intron XP_016880732.1
XM_017025244.1 Intron XP_016880733.1
XM_017025245.1 Intron XP_016880734.1

View Full Product Details