Product Details

SNP ID
rs8095307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:26141673 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAAAGAACAATAAGAACAAAAG[C/T]GTAGTTCTTTTTTTTCTTTTTTCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMA8 PubMed Links

Gene Details

Gene
PSMA8
Gene Name
proteasome subunit alpha 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025096.1 Intron NP_001020267.1
NM_001025097.1 Intron NP_001020268.1
NM_001308188.1 Intron NP_001295117.1
NM_144662.2 Intron NP_653263.2
XM_011525818.1 Intron XP_011524120.1

View Full Product Details